Skip to Content
Merck
All Photos(1)

Key Documents

280968

Sigma-Aldrich

N-(2-Mercaptopropionyl)glycine

99%

Synonym(s):

Tiopronin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(SH)CONHCH2COOH
CAS Number:
Molecular Weight:
163.19
Beilstein:
1859822
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Assay

99%

SMILES string

CC(S)C(=O)NCC(O)=O

InChI

1S/C5H9NO3S/c1-3(10)5(9)6-2-4(7)8/h3,10H,2H2,1H3,(H,6,9)(H,7,8)

InChI key

YTGJWQPHMWSCST-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaoqing Cai et al.
Analytical and bioanalytical chemistry, 401(3), 809-816 (2011-05-04)
Well-dispersed gold nanoparticles (NP) coated with tiopronin were synthesized by X-ray irradiation without reducing agents. High-resolution transmission electron microscopy shows that the average core diameters of the NPs can be systematically controlled by adjusting the tiopronin to Au mole ratio
Mildred Quintana et al.
Chemical communications (Cambridge, England), 48(100), 12159-12161 (2012-10-24)
Under ultrasonication, the production of high quality graphene layers by exfoliation of graphite was achieved via addition of tiopronin as an antioxidant.
Self-assembled ultra-high aspect ratio silver nanochains.
Matthew J Shiers et al.
Advanced materials (Deerfield Beach, Fla.), 24(38), 5227-5235 (2012-08-07)
Andrew S Goldsborough et al.
Journal of medicinal chemistry, 54(14), 4987-4997 (2011-06-11)
A major challenge in the treatment of cancer is multidrug resistance (MDR) that develops during chemotherapy. Here we demonstrate that tiopronin (1), a thiol-substituted N-propanoylglycine derivative, was selectively toxic to a series of cell lines expressing the drug efflux pump
Lei Cui et al.
Nanomedicine : nanotechnology, biology, and medicine, 9(2), 264-273 (2012-06-13)
The present study characterized the in vitro biological response of a comprehensive set of cancer cell lines to gold nanoparticles (2.7 nm) coated with tiopronin (AuNPs-TP). Our findings suggest that upon entering cells, the AuNPs-TP are sequestered in vacuoles such

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service