Skip to Content
Merck
All Photos(2)

Key Documents

280429

Sigma-Aldrich

(Triphenylphosphoranylidene)acetonitrile

97%

Synonym(s):

(Cyano)(triphenylphosphoranylidene)methane, (Cyanomethylene)triphenylphosphorane, Cyanomethyltriphenylphosphonium ylide, NSC 135204

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$154.00

$154.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$154.00

About This Item

Linear Formula:
(C6H5)3P=CHCN
CAS Number:
Molecular Weight:
301.32
EC Number:
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

$154.00


Check Cart for Availability

Request a Bulk Order

Assay

97%

reaction suitability

reaction type: C-C Bond Formation

mp

189-195 °C (lit.)

functional group

nitrile
phosphine

SMILES string

N#CC=P(c1ccccc1)(c2ccccc2)c3ccccc3

InChI

1S/C20H16NP/c21-16-17-22(18-10-4-1-5-11-18,19-12-6-2-7-13-19)20-14-8-3-9-15-20/h1-15,17H

InChI key

APISVOVOJVZIBA-UHFFFAOYSA-N

Application

Reactant for
  • Preparation of antitrypanosomal activity
  • Direct Michael addition of alkenes
  • Synthesis of 1,6-enynes
  • Beta-umpolung addition of nucleophiles to activated disubstituted alkyl allenes
  • Stereoselective analysis of β-homoproline derivatives
  • Stereoselective olefination of N-sulfonyl imines
  • Palladacycle mediated synthesis of cyano-functionalized chiral diphosphines
  • Horner-Wadsworth-Emmons reactions
Wittig reagent for the preparation of α,β, unsaturated nitriles from carbonyl compounds.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hiroaki Ohno et al.
The Journal of organic chemistry, 72(12), 4378-4389 (2007-05-11)
Thermal [2 + 2] cycloaddition of allenes with an additional multiple bond is described. By simply heating the allenenes or allenynes having a three-atom tether in an appropriate solvent such as dioxane or DMF, the distal double bond of the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service