Skip to Content
Merck
All Photos(2)

Key Documents

280003

Sigma-Aldrich

3,4-Dichlorophenylacetic acid

98%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$40.50
25 G
$136.00

$40.50


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$40.50
25 G
$136.00

About This Item

Linear Formula:
Cl2C6H3CH2CO2H
CAS Number:
Molecular Weight:
205.04
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$40.50


Check Cart for Availability

Request a Bulk Order

Assay

98%

form

solid

mp

89-91 °C (lit.)

functional group

carboxylic acid
chloro

SMILES string

OC(=O)Cc1ccc(Cl)c(Cl)c1

InChI

1S/C8H6Cl2O2/c9-6-2-1-5(3-7(6)10)4-8(11)12/h1-3H,4H2,(H,11,12)

InChI key

ZOUPGSMSNQLUNW-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B R de Costa et al.
Journal of medicinal chemistry, 32(8), 1996-2002 (1989-08-01)
The synthesis and in vitro sigma receptor activity of the two diastereomers of U50,488 [(+/-)-2], namely, (1R,2S)-(+)- cis-3,4-dichloro-N-methyl-N-[2-(1-pyrrolidinyl)cyclohexyl]benzeneacet ami de [(+)-1] and (1S,2R)-(-)-cis-3,4-dichloro- N-methyl-N-[2-(1-pyrrolidinyl)cyclohexyl]benzeneacetamide [(-)-1], are described. (+)-1 and (-)-1 were synthesized from (+/-)-trans-N-methyl-2-aminocyclohexanol [(+/-)-3]. Pyridinium chlorochromate (PCC) oxidation of
A Naylor et al.
Journal of medicinal chemistry, 37(14), 2138-2144 (1994-07-08)
The synthesis of 5-(arylacetyl)-4-[(alkylamino)methyl]furo[3,2-c] pyridines (16-23, 26, 27) and their activities as kappa-opioid receptor agonists are described. kappa-Agonist potency was particularly sensitive to the nature of the basic moiety. In particular, in the rabbit vas deferens (kappa-specific tissue), the 3-pyrrolidinol

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service