Skip to Content
Merck
All Photos(1)

Key Documents

279692

Sigma-Aldrich

Ethyl isothiocyanatoacetate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SCNCH2CO2C2H5
CAS Number:
Molecular Weight:
145.18
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.502 (lit.)

bp

104-106 °C/7 mmHg (lit.)

density

1.171 g/mL at 25 °C (lit.)

functional group

amine
ester
isothiocyanate

storage temp.

2-8°C

SMILES string

CCOC(=O)CN=C=S

InChI

1S/C5H7NO2S/c1-2-8-5(7)3-6-4-9/h2-3H2,1H3

InChI key

IYPSSPPKMLXXRN-UHFFFAOYSA-N

Application

Ethyl isothiocyanatoacetate has been used in the synthesis of:
  • fused pyrimidines
  • ethyl (3-substituted 5-thioxo-1,2,4-triazolin-4-yl)acetates via addition-cyclization reaction with carboxylic acid hydrazides in the presence of sodium ethoxide
  • 3-O-amino-2-thiohydantoins
  • quinazoline, benzothienopyrimidine and benzofuropyrimidine
  • thiazolo[5,4-d]thiazole

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

228.2 °F - closed cup

Flash Point(C)

109 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Improved access to thiazolo [5, 4-d] thiazole and thieno [2, 3-d] thiazole.
Rossler A and Boldt P.
Journal of the Chemical Society. Perkin Transactions 1, 4, 685-688 (1998)
A Z Chowdhury et al.
Chemical & pharmaceutical bulletin, 49(4), 391-395 (2001-04-20)
o-Aminonitrile or o-aminoester compounds were cyclized to fused pyrimidines by reacting with ethyl iso(thio)cyanatoacetate in pyridine, and then were methylated, halogenated and subsequently displaced by the amines studied.
Synthesis of 3-O-amino-2-thiohydantoins.
Ryczek J.
Journal of Heterocyclic Chemistry, 40(4), 665-670 (2003)
Addition-cyclization reactions of ethyl isothiocyanatoacetate with carboxylic acid hydrazides.
Veverka M and Marchalin M.
Collection of Czechoslovak Chemical Communications, 52(1), 113-119 (1987)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service