Skip to Content
Merck
All Photos(2)

Key Documents

279501

Sigma-Aldrich

1-Ethyl-4-piperidone

98%

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$120.00
100 G
$319.00

$120.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$120.00
100 G
$319.00

About This Item

Empirical Formula (Hill Notation):
C7H13NO
CAS Number:
Molecular Weight:
127.18
Beilstein:
107533
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$120.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.464 (lit.)

bp

46-48 °C/1 mmHg (lit.)

density

0.944 g/mL at 25 °C (lit.)

functional group

ketone

storage temp.

2-8°C

SMILES string

CCN1CCC(=O)CC1

InChI

1S/C7H13NO/c1-2-8-5-3-7(9)4-6-8/h2-6H2,1H3

InChI key

BDVKAMAALQXGLM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1-Ethyl-4-piperidone has been used in the preparation of 3,5-bisarylmethylene-1-ethyl-4-piperidones[1].

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

154.4 °F - closed cup

Flash Point(C)

68 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and anti-cancer activity of pyridine and thiazolopyrimidine derivatives using 1-ethylpiperidone as a synthon.
Hammam AE-FG, et al.
Indian J. Chem. B, 40(3), 213-221 (2001)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service