Skip to Content
Merck
All Photos(1)

Key Documents

278939

Sigma-Aldrich

Zinc selenite

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ZnSeO3
CAS Number:
Molecular Weight:
192.35
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
grade:
for analytical purposes
form:
powder

grade

for analytical purposes

form

powder

SMILES string

[Zn++].[O-][Se]([O-])=O

InChI

1S/H2O3Se.Zn/c1-4(2)3;/h(H2,1,2,3);/q;+2/p-2

InChI key

PEUPCBAALXHYHP-UHFFFAOYSA-L

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Da-Ke Shen et al.
Journal of Zhejiang University. Science, 4(2), 131-135 (2003-03-28)
MBE growth of ZnS(x)Se(1 - x) thin films on ITO coated glass substrates were carried out using ZnS and Se sources with the substrate temperature ranging from 270 degrees C to 330 degrees C . The XRD theta/2theta spectra resulted
R A Velázquez et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 19(6), 2288-2300 (1999-03-05)
Zinc appears to play a role in synaptic transmission in the hippocampus. We tested the hypothesis that zinc is similarly involved in sensory transmission by determining whether vesicular zinc and metallothionein-III (MT-III), a zinc-binding protein, are localized in rat primary
Seung Mook Jo et al.
Biometals : an international journal on the role of metal ions in biology, biochemistry, and medicine, 21(2), 151-158 (2007-06-16)
Zinc enriched (ZEN) neurons and terminals are abundant in the rodent spinal cord. Zinc ions have been suggested to modulate the excitability of primary afferent fibers believed to be important in nociceptive transmission. To test the hypothesis that vesicular zinc
Amelie Tsoutsoulopoulos et al.
Toxicology in vitro : an international journal published in association with BIBRA, 58, 245-255 (2019-03-21)
The CULTEX® Radial Flow System (RFS) is a modular in vitro system for the homogenous exposure of cells to airborne particles at the air-liquid interface (ALI). A former pre-validation study successfully demonstrated the general applicability of the CULTEX® RFS and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service