Skip to Content
Merck
All Photos(1)

Key Documents

277975

Sigma-Aldrich

Butyldiisopropoxyborane

98%

Synonym(s):

Diisopropyl butylboronate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3B[OCH(CH3)2]2
CAS Number:
Molecular Weight:
186.10
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Assay

98%

form

liquid

refractive index

n20/D 1.395 (lit.)

bp

145-147 °C/746 mmHg (lit.)

density

0.794 g/mL at 25 °C (lit.)

SMILES string

CCCCB(OC(C)C)OC(C)C

InChI

1S/C10H23BO2/c1-6-7-8-11(12-9(2)3)13-10(4)5/h9-10H,6-8H2,1-5H3

InChI key

QNRNUKUCUSOJKW-UHFFFAOYSA-N

Application

Potential derivatizing agent for difunctional compounds for GC and GC-MS analysis.[1]

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

84.2 °F - closed cup

Flash Point(C)

29 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aldrichimica Acta, 18, 80-80 (1985)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service