Skip to Content
Merck
All Photos(1)

Key Documents

277711

Sigma-Aldrich

(Methoxymethyl)trimethylsilane

98%

Synonym(s):

(Trimethylsilyl)methyl methyl ether

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$232.00

$232.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
$232.00

About This Item

Linear Formula:
CH3OCH2Si(CH3)3
CAS Number:
Molecular Weight:
118.25
Beilstein:
1732033
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

$232.00


Check Cart for Availability

Request a Bulk Order

Assay

98%

form

liquid

refractive index

n20/D 1.39 (lit.)

bp

83 °C/740 mmHg (lit.)

density

0.758 g/mL at 25 °C (lit.)

functional group

ether

SMILES string

COC[Si](C)(C)C

InChI

1S/C5H14OSi/c1-6-5-7(2,3)4/h5H2,1-4H3

InChI key

PPNQXAYCPNTVHH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

21.2 °F - closed cup

Flash Point(C)

-6 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Review SET mechanism in photoredox catalysis for sustainable reactions and rearrangement of oxaziridines.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service