Skip to Content
Merck
All Photos(2)

Documents

277398

Sigma-Aldrich

O-Isopropylxanthic acid potassium salt

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHOCS2K
CAS Number:
Molecular Weight:
174.33
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

264 °C (dec.) (lit.)

SMILES string

CC(C)OC(=S)S[K]

InChI

1S/C4H8OS2.K/c1-3(2)5-4(6)7;/h3H,1-2H3,(H,6,7);/q;+1/p-1

InChI key

ZMWBGRXFDPJFGC-UHFFFAOYSA-M

Related Categories

Application

O-Isopropylxanthic acid potassium salt may be used in chemical synthesis.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F Hissner et al.
Journal of chromatography. A, 853(1-2), 497-502 (1999-09-16)
Alkyl xanthates (O-alkyl dithiocarbonates) and phosphonates are important organic collectors for the flotation of metals from crude ore. Leaching from waste dumps into river and ground water, these substances can cause environmental pollution. A capillary electrophoretic method for the routine
V B Dorogova et al.
Gigiena truda i professional'nye zabolevaniia, (3)(3), 46-47 (1990-01-01)
The article contains a description of a photometric technique for xanthate detecting in the air in the working zone area. The detection technique is based on the xanthates' interaction with the nickel salts, with the formation of yellow--coloured nickel xanthates
M Alijanianzadeh et al.
Journal of enzyme inhibition and medicinal chemistry, 22(2), 239-246 (2007-05-24)
Three iso-alkyldithiocarbonates (xanthates), as sodium salts, C3H7OCS2Na (I), C4H9OCS2Na (II) and C5H11OCS2Na (III), were synthesized, by the reaction between CS2 with the corresponding iso-alcohol in the presence of NaOH, and examined for inhibition of both cresolase and catecholase activities of
M N Chandraprabha et al.
Journal of colloid and interface science, 276(2), 323-332 (2004-07-24)
Effective methods for selective separation using flotation or flocculation of arsenopyrite from pyrite by biomodulation using Acidithiobacillus ferrooxidans are presented here. Adhesion of the bacterium to the surface of arsenopyrite was very slow compared to that to pyrite, resulting in
Mohammad S M Saifullah et al.
ACS nano, 11(10), 9920-9929 (2017-09-25)
Nanostructures of metal sulfides are conventionally prepared via chemical techniques and patterned using self-assembly. This poses a considerable amount of challenge when arbitrary shapes and sizes of nanostructures are desired to be placed at precise locations. Here, we describe an

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service