Skip to Content
Merck
All Photos(1)

Key Documents

276324

Sigma-Aldrich

Tungsten

rod, diam. 4.0 mm, 99.99% trace metals basis

Synonym(s):

W

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
W
CAS Number:
Molecular Weight:
183.84
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

Assay

99.99% trace metals basis

form

rod

resistivity

4.9 μΩ-cm, 20°C

diam.

4.0 mm

bp

5660 °C (lit.)

mp

3410 °C (lit.)

density

19.3 g/mL at 25 °C (lit.)

SMILES string

[W]

InChI

1S/W

InChI key

WFKWXMTUELFFGS-UHFFFAOYSA-N

Quantity

24 g = 100 mm

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Weijie Zhao et al.
ACS nano, 7(1), 791-797 (2012-12-22)
Geometrical confinement effect in exfoliated sheets of layered materials leads to significant evolution of energy dispersion in mono- to few-layer thickness regime. Molybdenum disulfide (MoS(2)) was recently found to exhibit indirect-to-direct gap transition when the thickness is reduced to a
Karel Deprez et al.
Medical physics, 40(1), 012501-012501 (2013-01-10)
The construction of complex collimators with a high number of oblique pinholes is very labor intensive, expensive or is sometimes impossible with the current available techniques (drilling, milling or electric discharge machining). All these techniques are subtractive: one starts from
Zhigang Chen et al.
Advanced materials (Deerfield Beach, Fla.), 25(14), 2095-2100 (2013-02-22)
A new photothermal coupling agent for photothermal ablation (PTA) therapy of tumors is developed based on ultrathin PEGylated W18O49 nanowires. After being injected with the nanowire solution, the in vivo tumors exhibit a rapid temperature rise to 50.0 ± 0.5
Denys Shevchenko et al.
Dalton transactions (Cambridge, England : 2003), 42(14), 5130-5139 (2013-02-14)
A rare example of a "monomeric" triple transition-metal substituted Keggin anion has been synthesized and characterized by various methods including X-ray crystallography, ESI and MALDI mass spectrometry, electrochemistry, EPR, and SQUID.
Michael L Crichton et al.
Biomaterials, 34(8), 2087-2097 (2012-12-25)
The recent emergence of micro-devices for vaccine delivery into upper layers of the skin holds potential for increased immune responses using physical means to target abundant immune cell populations. A challenge in doing this has been a limited understanding of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service