Skip to Content
Merck
All Photos(1)

Key Documents

276154

Sigma-Aldrich

Methyl 3-amino-2-pyrazinecarboxylate

97%

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$100.00

$100.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
$100.00

About This Item

Empirical Formula (Hill Notation):
C6H7N3O2
CAS Number:
Molecular Weight:
153.14
Beilstein:
127495
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$100.00


Check Cart for Availability

Request a Bulk Order

Assay

97%

mp

169-172 °C (lit.)

SMILES string

COC(=O)c1nccnc1N

InChI

1S/C6H7N3O2/c1-11-6(10)4-5(7)9-3-2-8-4/h2-3H,1H3,(H2,7,9)

InChI key

INCSQLZZXBPATR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Methyl 3-amino-2-pyrazinecarboxylate has been used in:
  • synthesis of 2-arylpteridin-4-ones[1] and piperazine-derived 2-furan-2-yl-[1,2,4]triazolo[1,5-a] pyrazines[2]
  • reactions with isocyanates[3] and aroyl chlorides[1] for conversion to pteridinediones

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

First synthesis of piperazine-derived [1, 2, 4] triazolo [1, 5-a] pyrazine as an adenosine A2a receptor antagonist.
Peng H, et al.
Heterocycles, 65(10), 2321-2327 (2005)
Dihalogentriphenylphosphorane in der Heterocyclensynthese, 29. Eine einfache Synthese von Pteridin-4-onen aus 3-Amino-2-pyrazincarbonsauremethylester und Pyrazino [3, 1] oxazin-4-onen.
Wamhoff H and Kroth E.
Synthesis, 04, 405-410 (1994)
Heterocycles, 35, 949-949 (1993)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service