Skip to Content
Merck
All Photos(1)

Key Documents

269255

Sigma-Aldrich

trans-2-Pentenal

95%

Synonym(s):

2-Ethylacrylic aldehyde

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$69.40
25 G
$184.00

$69.40


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$69.40
25 G
$184.00

About This Item

Linear Formula:
C2H5CH=CHCHO
CAS Number:
Molecular Weight:
84.12
Beilstein:
1719742
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$69.40


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

95%

refractive index

n20/D 1.440-1.446 (lit.)

bp

80-81 °C/160 mmHg (lit.)

density

0.86 g/mL at 25 °C (lit.)

functional group

aldehyde

SMILES string

CC\C=C\C=O

InChI

1S/C5H8O/c1-2-3-4-5-6/h3-5H,2H2,1H3/b4-3+

InChI key

DTCCTIQRPGSLPT-ONEGZZNKSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Trans-2-Pentenal (pentenal) is an α,β-unsaturated aldehyde that increases [Ca2+] in cultured neonatal rat trigeminal ganglion (TG) neurons[1]. The gas-phase reactions of NO3 radicals with pentenal in pure N2, using a relative rate technique, was studied[2].

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

71.6 °F - closed cup

Flash Point(C)

22 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhijun Zhao et al.
The journal of physical chemistry. A, 115(44), 12161-12172 (2011-10-15)
The gas-phase reactions of NO(3) radicals with series of 1-alkenes, dienes, cycloalkenes, alkenols, and alkenals were studied in pure N(2) or 20% O(2)/80% N(2) bath gas at room temperature and atmospheric pressure using a relative rates technique. Rate coefficients were
Takashi Inoue et al.
Cellular and molecular neurobiology, 30(1), 35-41 (2009-07-02)
Trans-2-Pentenal (pentenal), an alpha,beta-unsaturated aldehyde, induces increases in [Ca(2+)](i) in cultured neonatal rat trigeminal ganglion (TG) neurons. Since all pentenal-sensitive neurons responded to a specific TRPA1 agonist, allyl isothiocyanate (AITC) and neurons from TRPA1 knockouts failed to respond to pentenal
M Grootveld et al.
The Journal of clinical investigation, 101(6), 1210-1218 (1998-04-29)
Thermal stressing of polyunsaturated fatty acid (PUFA)- rich culinary oils according to routine frying or cooking practices generates high levels of cytotoxic aldehydic products (predominantly trans-2-alkenals, trans,trans-alka-2,4-dienals, cis,trans-alka-2, 4-dienals, and n-alkanals), species arising from the fragmentation of conjugated hydroperoxydiene precursors.
M E Davis et al.
Physical chemistry chemical physics : PCCP, 9(18), 2240-2248 (2007-05-10)
Rate coefficients for the gas-phase reaction of the OH radical with (E)-2-pentenal (CH(3)CH(2)CH[double bond]CHCHO), (E)-2-hexenal (CH(3)(CH(2))(2)CH[double bond]CHCHO), and (E)-2-heptenal (CH(3)(CH(2))(3)CH[double bond]CHCHO), a series of unsaturated aldehydes, over the temperature range 244-374 K at pressures between 23 and 150 Torr (He
R Canonero et al.
Mutation research, 244(2), 153-156 (1990-06-01)
Five alk-2-enals--pent-2-enal, hex-2-enal, hept-2-enal, oct-2-enal and non-2-enal--produced by lipid peroxidation were tested for mutagenic activity in V79 Chinese hamster cells. At concentrations ranging from 0.003 to 0.3 mM all 5 alk-2-enals induced a dose-dependent increase in the frequency of 6-thioguanine-resistant

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service