Skip to Content
Merck
All Photos(1)

Key Documents

264377

Sigma-Aldrich

2,4-Difluorobenzylamine

98%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$124.00

$124.00


0 left in stock

Request a Bulk Order

Select a Size

Change View
5 G
$124.00

About This Item

Linear Formula:
F2C6H3CH2NH2
CAS Number:
Molecular Weight:
143.13
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$124.00


0 left in stock

Request a Bulk Order

Assay

98%

form

liquid

refractive index

n20/D 1.49 (lit.)

density

1.204 g/mL at 25 °C (lit.)

functional group

amine
fluoro

SMILES string

NCc1ccc(F)cc1F

InChI

1S/C7H7F2N/c8-6-2-1-5(4-10)7(9)3-6/h1-3H,4,10H2

InChI key

QDZZDVQGBKTLHV-UHFFFAOYSA-N

Application

2,4-Difluorobenzylamine has been used in the synthesis of βAla–Aib(N-fluoroarylmethyl)], nonpolar nucleobase dipeptide via Ugi four-component condensation reaction[1].

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

154.4 °F - closed cup

Flash Point(C)

68 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Design and synthesis of N-nonpolar nucleobase dipeptides: application of the Ugi reaction for the preparation of dipeptides having fluoroarylalkyl groups appended to the nitrogen atom.
Das BK, et al.
Journal of the Chemical Society. Perkin Transactions 1, 2, 197-206 (2002)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service