Skip to Content
Merck
All Photos(2)

Key Documents

263710

Sigma-Aldrich

4-(Methylamino)butyric acid hydrochloride

99%

Synonym(s):

4-(Methylamino)butanoic acid hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$79.40
100 G
$198.00

$79.40


Estimated to ship on14 April 2025


Request a Bulk Order

Select a Size

Change View
25 G
$79.40
100 G
$198.00

About This Item

Linear Formula:
CH3NH(CH2)3CO2H · HCl
CAS Number:
Molecular Weight:
153.61
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

$79.40


Estimated to ship on14 April 2025


Request a Bulk Order

Assay

99%

reaction suitability

reaction type: solution phase peptide synthesis

mp

124-126 °C (lit.)

application(s)

peptide synthesis

SMILES string

Cl[H].CNCCCC(O)=O

InChI

1S/C5H11NO2.ClH/c1-6-4-2-3-5(7)8;/h6H,2-4H2,1H3,(H,7,8);1H

InChI key

JMLHQRQZCMCQNJ-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Prashanth Srinivasan et al.
Nature communications, 10(1), 3634-3634 (2019-08-14)
Tropane alkaloids (TAs) are a class of phytochemicals produced by plants of the nightshade family used for treating diverse neurological disorders. Here, we demonstrate de novo production of tropine, a key intermediate in the biosynthetic pathway of medicinal TAs such

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service