Skip to Content
Merck
All Photos(1)

Key Documents

263176

Sigma-Aldrich

Praseodymium

powder, ~40 mesh, 99.5% trace rare earth metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Pr
CAS Number:
Molecular Weight:
140.91
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.23

Assay

99.5% trace rare earth metals basis

form

powder

reaction suitability

reagent type: catalyst
core: praseodymium

resistivity

68 μΩ-cm, 20°C

particle size

~40 mesh

bp

3520 °C (lit.)

mp

931 °C (lit.)

density

6.71 g/mL at 25 °C (lit.)

SMILES string

[Pr]

InChI

1S/Pr

InChI key

PUDIUYLPXJFUGB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 4 - Eye Irrit. 2 - Pyr. Sol. 1 - Skin Irrit. 2

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Junichiro Kojou et al.
Applied optics, 51(9), 1382-1386 (2012-03-24)
We demonstrate pulse laser operation of a Pr:LiYF(4) laser pumped by InGaN laser diodes (444 nm) using an acousto-optic modulator. We obtained a maximum laser peak power of 167 W (4 μJ/pulse) with a pulse width of 24 ns at
Liaolin Zhang et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 93, 223-227 (2012-04-10)
We report on the spectroscopic properties of Pr(3+)-doped boro-phosphate, boro-germo-silicate and tellurite glasses. The stimulated absorption and emission cross sections were estimated. Only one emission at 596 nm and 605 nm is observed in Pr(3+)-doped boro-phosphate and boro-germo-silicate glasses, respectively
N Bendangsenla et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 103, 160-166 (2012-12-22)
The interactions of Pr(III) with nucleosides and nucleotides have been studied in different organic solvents employing absorption difference and comparative absorption spectrophotometry. The magnitudes of the variations in both energy and intensity interaction parameters were used to explore the degree
Mozhgan Khorasani-Motlagh et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 79(5), 978-984 (2011-06-15)
The binding of neodymium(III) and praseodymium(III) complexes containing 1,10-phenanthroline, [M(phen)2Cl3·OH2] (M=Nd (1), Pr (2)), to DNA has been investigated by absorption, emission, and viscosity measurements. The complexes show absorption decreasing in charge transfer band, fluorescence decrement when bound to DNA.
D V Sunitha et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 99, 279-287 (2012-10-23)
A series of Pr(3+) (1-9 mol%) doped CdSiO(3) nanophosphors have been prepared for the first time by a low temperature solution combustion method using oxalyldihydrizide (ODH) as a fuel. The final product was characterized by Powder X-ray diffraction (PXRD), Fourier

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service