Skip to Content
Merck
All Photos(2)

Documents

261130

Sigma-Aldrich

Lanthanum

ingot (under oil), 99.9% trace rare earth metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
La
CAS Number:
Molecular Weight:
138.91
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

Assay

99.9% trace rare earth metals basis

form

ingot (under oil)

reaction suitability

reagent type: catalyst
core: lanthanum

resistivity

54 μΩ-cm

bp

3464 °C (lit.)

mp

920 °C (lit.)

density

6.19 g/mL at 25 °C (lit.)

SMILES string

[La]

InChI

1S/La

InChI key

FZLIPJUXYLNCLC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Water-react 1

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sebastian Meis et al.
Water research, 47(13), 4460-4473 (2013-06-15)
Phoslock(®) is increasingly used worldwide to control sediment phosphorus (P) release and cyanobacterial blooms despite the fact that little is known about its mode of action in lake bed sediments. This study quantified the effects of Phoslock(®) on sediment elemental
Mark F McLaughlin et al.
PloS one, 8(1), e54531-e54531 (2013-01-26)
Targeted radiotherapies maximize cytotoxicty to cancer cells. In vivo α-generator targeted radiotherapies can deliver multiple α particles to a receptor site dramatically amplifying the radiation dose delivered to the target. The major challenge with α-generator radiotherapies is that traditional chelating
F Borodavka et al.
Journal of physics. Condensed matter : an Institute of Physics journal, 25(8), 085901-085901 (2013-01-31)
The lattice dynamics of La(1/2)Na(1/2)TiO(3) single crystal have been investigated by far-infrared and Raman scattering spectroscopy. Far-IR reflectivity spectra revealed Slater, Last and Axe modes near 140, 190 and 555 cm(-1), respectively. The frequency of the Slater mode decreases by about
Yin-zhu Wang et al.
Biosensors & bioelectronics, 48, 56-60 (2013-05-08)
In this paper, a nonenzymatic glucose biosensor based on perovskite LaTiO3-Ag0.2(LTA) modified electrode was presented. The morphology and the composition of the perovskite LaTiO₃-Ag0.2 nanomaterials were characterized by using scanning electron microscopy (SEM) and X-ray diffraction (XRD) respectively. The LaTiO₃-Ag0.2(LTA)
Kasper Reitzel et al.
Water research, 47(8), 2787-2796 (2013-03-26)
Effects of pH, alkalinity and conductivity on the adsorption of soluble reactive phosphorus (SRP) onto lanthanum (La) modified bentonite clay (Phoslock(®)) were investigated in laboratory experiments using eight different types of filtered water representing freshwater with low and normal alkalinity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service