Skip to Content
Merck
All Photos(1)

Documents

261106

Sigma-Aldrich

Europium

chunks, under oil, purified by distillation, 99.9%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Eu
CAS Number:
Molecular Weight:
151.96
EC Number:
MDL number:
UNSPSC Code:
12352300

Assay

99.9%

form

chunks

purified by

distillation

resistivity

81 μΩ-cm, 20°C

bp

1527 °C (lit.)

mp

822 °C (lit.)

density

5.24 g/mL at 25 °C (lit.)

SMILES string

[Eu]

InChI

1S/Eu

InChI key

OGPBJKLSAFTDLK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Pyr. Sol. 1

Supplementary Hazards

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service