Skip to Content
Merck
All Photos(1)

Key Documents

260266

Sigma-Aldrich

1,2-Epoxytetradecane

technical grade, 85%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H28O
CAS Number:
Molecular Weight:
212.37
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

grade

technical grade

Assay

85%

form

liquid

refractive index

n20/D 1.4408 (lit.)

bp

95-96 °C/0.4 mmHg (lit.)

density

0.845 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCCCCC1CO1

InChI

1S/C14H28O/c1-2-3-4-5-6-7-8-9-10-11-12-14-13-15-14/h14H,2-13H2,1H3

InChI key

IOHJQSFEAYDZGF-UHFFFAOYSA-N

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

221.0 °F - closed cup

Flash Point(C)

105 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ping Song et al.
Molecular therapy : the journal of the American Society of Gene Therapy, 27(8), 1424-1435 (2019-06-04)
Interleukin-1 beta (IL-1β) plays a central role in the induction of rheumatoid arthritis (RA). In the present study, we demonstrated that lipidoid-polymer hybrid nanoparticle (FS14-NP) can efficiently deliver siRNA against IL-1β (siIL-1β) to macrophages and effectively suppress the pathogenesis of
Shantipriya Khadka et al.
Electrophoresis, 37(23-24), 3160-3171 (2016-09-10)
A novel precursor monolithic capillary column referred to as "hydroxy monolith" or OHM was prepared by the in situ copolymerization of hydroxyethylmethacrylate (HEMA) with pentaerythritol triacrylate (PETA) yielding the neutral poly(HEMA-co-PETA) monolith. The neutral precursor OHM capillary thus obtained was
Eva Ramsay et al.
Pharmaceutics, 12(7) (2020-07-28)
Inflammation is involved in the pathogenesis of several age-related ocular diseases, such as macular degeneration (AMD), diabetic retinopathy, and glaucoma. The delivery of anti-inflammatory siRNA to the retinal pigment epithelium (RPE) may become a promising therapeutic option for the treatment
Wei Tao et al.
Science translational medicine, 12(553) (2020-07-29)
Atherosclerotic lesional macrophages express molecules that promote plaque progression, but lack of mechanisms to therapeutically target these molecules represents a major gap in translational cardiovascular research. Here, we tested the efficacy of a small interfering RNA (siRNA) nanoparticle (NP) platform
Rachel S Riley et al.
Science advances, 7(3) (2021-02-02)
Clinical advances enable the prenatal diagnosis of genetic diseases that are candidates for gene and enzyme therapies such as messenger RNA (mRNA)-mediated protein replacement. Prenatal mRNA therapies can treat disease before the onset of irreversible pathology with high therapeutic efficacy

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service