Skip to Content
Merck
All Photos(1)

Documents

258490

Sigma-Aldrich

N6-Benzoyl-5′-O-(4,4′-dimethoxytrityl)-2′-deoxyadenosine

98%

Synonym(s):

Bz-DMT-dA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C38H35N5O6
CAS Number:
Molecular Weight:
657.71
Beilstein:
602310
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

98%

SMILES string

COc1ccc(cc1)C(OC[C@H]2O[C@H](C[C@@H]2O)n3cnc4c(NC(=O)c5ccccc5)ncnc34)(c6ccccc6)c7ccc(OC)cc7

InChI

1S/C38H35N5O6/c1-46-29-17-13-27(14-18-29)38(26-11-7-4-8-12-26,28-15-19-30(47-2)20-16-28)48-22-32-31(44)21-33(49-32)43-24-41-34-35(39-23-40-36(34)43)42-37(45)25-9-5-3-6-10-25/h3-20,23-24,31-33,44H,21-22H2,1-2H3,(H,39,40,42,45)/t31-,32+,33+/m0/s1

InChI key

LPICNYATEWGYHI-WIHCDAFUSA-N

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service