Skip to Content
Merck
All Photos(1)

Key Documents

257605

Sigma-Aldrich

2,3,7,8,12,13,17,18-Octaethyl-21H,23H-porphine cobalt(II)

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$167.00

$167.00


Estimated to ship on29 March 2025


Request a Bulk Order

Select a Size

Change View
100 MG
$167.00

About This Item

Empirical Formula (Hill Notation):
C36H44CoN4
CAS Number:
Molecular Weight:
591.69
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

$167.00


Estimated to ship on29 March 2025


Request a Bulk Order

form

solid

Quality Level

λmax

393 nm

SMILES string

CCc1c(CC)c2cc3c(CC)c(CC)c4cc5nc(cc6c(CC)c(CC)c(cc1n2)n6[Co]n34)c(CC)c5CC

InChI

1S/C36H44N4.Co/c1-9-21-22(10-2)30-18-32-25(13-5)26(14-6)34(39-32)20-36-28(16-8)27(15-7)35(40-36)19-33-24(12-4)23(11-3)31(38-33)17-29(21)37-30;/h17-20H,9-16H2,1-8H3;/q-2;+2/b29-17-,30-18-,31-17-,32-18-,33-19-,34-20-,35-19-,36-20-;

InChI key

CILRATIHXSICFS-XTPDIVBZSA-N

General description

2,3,7,8,12,13,17,18-Octaethyl-21H,23H-porphine metalates with cobalt(II) to form 2,3,7,8,12,13,17,18-Octaethyl-21H,23H-porphine cobalt(II) (CoOEP). CoOEP may be used to form Langmuir Blodgett (LB) thin films.1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service