Skip to Content
Merck
All Photos(3)

Documents

256277

Sigma-Aldrich

(R,R)-(+)-Hydrobenzoin

99%, optical purity ee: 99% (HPLC)

Synonym(s):

(R,R)-1,2-Diphenyl-1,2-ethanediol, (R,R)-1,2-Diphenylethylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH(OH)CH(OH)C6H5
CAS Number:
Molecular Weight:
214.26
Beilstein:
2050815
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

optical activity

[α]28/D +93°, c = 2.5 in ethanol

optical purity

ee: 99% (HPLC)

mp

146-149 °C (lit.)

SMILES string

O[C@@H]([C@H](O)c1ccccc1)c2ccccc2

InChI

1S/C14H14O2/c15-13(11-7-3-1-4-8-11)14(16)12-9-5-2-6-10-12/h1-10,13-16H/t13-,14-/m1/s1

InChI key

IHPDTPWNFBQHEB-ZIAGYGMSSA-N

Looking for similar products? Visit Product Comparison Guide

General description

(R,R)-(+)-Hydrobenzoin is a chiral 1,2-diol useful as a chiral reagent, building block, ligand or auxiliary in asymmetric synthesis.

Application

(R,R)-(+)-Hydrobenzoin may be used in the oxyselenenylation step in the multi-step synthesis of cyclopentitols and aminocyclopentitols from cyclopentene. It may also be used as a ligand for the asymmetric addition of diethylzinc to aldehydes in the presence or absence of titanium tetra-isopropoxide to form (R)- or (S)-form of the corresponding secondary alcohol, respectively.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Divergent enantioselective pathways in the catalytic asymmetric addition of diethylzinc to aldehydes in the presence and absence of titanium tetraisopropoxide.
Dean MA and Hitchcock SR.
Tetrahedron Asymmetry, 19(22), 2563-2567 (2008)
Synthesis of enantiopure cyclopentitols and aminocyclopentitols mediated by oxyselenenylation of cyclopentene with (R, R)-hydrobenzoin.
Kim KS, et al.
Tetrahedron Letters, 39(36), 6471-6474 (1998)
A practical stereoselective synthesis of chiral hydrobenzoins via asymmetric transfer hydrogenation of benzils.
Murata K, et al.
Organic Letters, 1(7), 1119-1121 (1999)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service