Skip to Content
Merck
All Photos(1)

Key Documents

254525

Sigma-Aldrich

tert-Butyl propionate

99%

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$168.00
100 G
$301.00

$168.00


Available to ship on22 April 2025Details


Request a Bulk Order

Select a Size

Change View
25 G
$168.00
100 G
$301.00

About This Item

Linear Formula:
CH3CH2COOC(CH3)3
CAS Number:
Molecular Weight:
130.18
Beilstein:
1747857
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$168.00


Available to ship on22 April 2025Details


Request a Bulk Order

Assay

99%

form

liquid

refractive index

n20/D 1.393 (lit.)

bp

118-118.5 °C (lit.)

density

0.865 g/mL at 25 °C (lit.)

functional group

ester

SMILES string

CCC(=O)OC(C)(C)C

InChI

1S/C7H14O2/c1-5-6(8)9-7(2,3)4/h5H2,1-4H3

InChI key

JAELLLITIZHOGQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Stereochemistry of kinetic Michael addition of the Z and E lithium enolates of tert-butyl propionate has been investigated[1].

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

69.8 °F - closed cup

Flash Point(C)

21 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Acyclic stereoselection. 31. Stereoselection in the Michael addition reaction. 3. Relationship between ester enolate geometry and adduct stereochemistry in the kinetic Michael reaction of lithium enolates with enones.
Heathcock CH and Oare DA.
The Journal of Organic Chemistry, 50(16), 3022-3024 (1985)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service