253057
Ethyl m-toluate
99%
Synonym(s):
3-Methylbenzoic acid ethyl ester
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CH3C6H4CO2C2H5
CAS Number:
Molecular Weight:
164.20
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
99%
form
liquid
refractive index
n20/D 1.506 (lit.)
bp
110 °C/20 mmHg (lit.)
density
1.03 g/mL at 25 °C (lit.)
SMILES string
CCOC(=O)c1cccc(C)c1
InChI
1S/C10H12O2/c1-3-12-10(11)9-6-4-5-8(2)7-9/h4-7H,3H2,1-2H3
InChI key
WSJNYOVBJSOQST-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Ethyl m-toluate may be used in chemical synthesis.
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
213.8 °F - closed cup
Flash Point(C)
101 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service