Skip to Content
Merck
All Photos(1)

Key Documents

252972

Sigma-Aldrich

Diphenylvinylphosphine

95%

Synonym(s):

Ethenyldiphenylphosphine, Vinyldiphenylphosphine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)2PCH=CH2
CAS Number:
Molecular Weight:
212.23
EC Number:
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

liquid

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand

refractive index

n20/D 1.626 (lit.)

bp

104 °C/0.25 mmHg (lit.)

density

1.067 g/mL at 25 °C (lit.)

functional group

phosphine

SMILES string

C=CP(c1ccccc1)c2ccccc2

InChI

1S/C14H13P/c1-2-15(13-9-5-3-6-10-13)14-11-7-4-8-12-14/h2-12H,1H2

InChI key

AJVBXLXLODZUME-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service