Skip to Content
Merck
All Photos(1)

Key Documents

252913

Sigma-Aldrich

5,10,15,20-Tetrakis(pentafluorophenyl)-21H,23H-porphyrin iron(III) chloride

≥95% (HPLC)

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$261.00

$261.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$261.00

About This Item

Empirical Formula (Hill Notation):
C44H8ClF20FeN4
CAS Number:
Molecular Weight:
1063.83
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

$261.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥95% (HPLC)

form

powder

reaction suitability

reagent type: catalyst
core: iron

λmax

415 nm
498 nm (2nd)

SMILES string

Fc1c(F)c(F)c(c(F)c1F)-c2c3ccc(n3)c(-c4c(F)c(F)c(F)c(F)c4F)c5ccc6c(-c7c(F)c(F)c(F)c(F)c7F)c8ccc(n8)c(-c9c(F)c(F)c(F)c(F)c9F)c%10ccc2n%10[Fe](Cl)n56

InChI

1S/C44H8F20N4.ClH.Fe/c45-25-21(26(46)34(54)41(61)33(25)53)17-9-1-2-10(65-9)18(22-27(47)35(55)42(62)36(56)28(22)48)12-5-6-14(67-12)20(24-31(51)39(59)44(64)40(60)32(24)52)16-8-7-15(68-16)19(13-4-3-11(17)66-13)23-29(49)37(57)43(63)38(58)30(23)50;;/h1-8H;1H;/q-2;;+3/p-1/b17-9+,17-11+,18-10+,18-12+,19-13+,19-15+,20-14+,20-16+;;

InChI key

RVNJZXLLVARDGN-VTVSBRCESA-M

Application

Catalyst for epoxidations with peroxides,[1] oxidative decarbonylations,[2] and aliphatic hydroxylations.[3]

Features and Benefits

Catalyst for epoxidations with peroxides, oxidative decarbonylations, and aliphatic hydroxylations.
Preferred catalyst in catalytic oxygenation reactions.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Journal of the American Chemical Society, 115, 2775-2775 (1993)
Journal of the American Chemical Society, 114, 1308-1308 (1992)
Tetrahedron Letters, 33, 4949-4949 (1992)

Articles

Novel Graphene‑Based Nanostructures Production, Functionalization, and Engineering

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service