Skip to Content
Merck
All Photos(1)

Key Documents

252328

Sigma-Aldrich

1,2-Dibromobutane

97%

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$386.00

$386.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$386.00

About This Item

Linear Formula:
C2H5CH(Br)CH2Br
CAS Number:
Molecular Weight:
215.91
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$386.00


Check Cart for Availability

Request a Bulk Order

Assay

97%

form

liquid

refractive index

n20/D 1.5141 (lit.)

bp

59-60 °C/20 mmHg (lit.)

solubility

alcohol: miscible(lit.)
water: insoluble(lit.)

density

1.789 g/mL at 25 °C (lit.)

functional group

bromo

SMILES string

CCC(Br)CBr

InChI

1S/C4H8Br2/c1-2-4(6)3-5/h4H,2-3H2,1H3

InChI key

CZWSZZHGSNZRMW-UHFFFAOYSA-N

General description

Mechanism of Co(II) (N,N′-disalicylidene-ethylenediamine) catalyzed reduction of 1,2-dibromobutane in DMF has been investigated by microelectrode voltammetry[1].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A microelectrode study of the catalysis of alkyl halide reduction by Co (II)(salen).
Pletcher D and Thompson H.
Journal of Electroanalytical Chemistry, 464(2), 168-175 (1999)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service