Skip to Content
Merck
All Photos(1)

Key Documents

251542

Sigma-Aldrich

Methyl (S)-(+)-mandelate

≥99%

Synonym(s):

(+)-Methyl (S)-α-hydroxyphenylacetate, (+)-Methyl L-mandelate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH(OH)CO2CH3
CAS Number:
Molecular Weight:
166.17
Beilstein:
2614741
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22
Assay:
≥99%

Assay

≥99%

optical activity

[α]20/D +144°, c = 1 in methanol

optical purity

ee: 98% (HPLC)

mp

56-58 °C (lit.)

functional group

ester
hydroxyl
phenyl

SMILES string

COC(=O)[C@@H](O)c1ccccc1

InChI

1S/C9H10O3/c1-12-9(11)8(10)7-5-3-2-4-6-7/h2-6,8,10H,1H3/t8-/m0/s1

InChI key

ITATYELQCJRCCK-QMMMGPOBSA-N

General description

Optically active building block.

Application

Methyl (S)-(+)-mandelate may be used in the preparation of (S)-(-)2-hydroxy-1,2,2-triphenylethyl acetate[1] and (α′S)-α′-methoxycarbonylbenzyl (2S,4R)-(+)-1,2,4,5-tetrahydro-4-methyl-7,8-methylenedioxy-5-oxo-3-benzothiepin-2-carboxylate.[2]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

(R)-(+)-2-Hydroxy-1, 2, 2-Triphenylethyl Acetate.
Braun M, et al.
Organic Syntheses, 32-32 (1993)
Synthesis of novel 2-benzothiopyran and 3-benzothiepin derivatives and their stimulatory effect on bone formation 1.
Oda T, et al.
Journal of Medicinal Chemistry, 42(4), 751-760 (1999)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service