Skip to Content
Merck
All Photos(1)

Key Documents

247707

Sigma-Aldrich

4-Chloro-2-fluorotoluene

≥99%

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$153.00

$153.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$153.00

About This Item

Linear Formula:
CH3C6H3(Cl)F
CAS Number:
Molecular Weight:
144.57
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$153.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥99%

form

liquid

refractive index

n20/D 1.498 (lit.)

bp

158 °C/743 mmHg (lit.)

density

1.186 g/mL at 25 °C (lit.)

functional group

chloro
fluoro

SMILES string

Cc1ccc(Cl)cc1F

InChI

1S/C7H6ClF/c1-5-2-3-6(8)4-7(5)9/h2-4H,1H3

InChI key

MKFCYQTVSDCXAQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Fourier Transform (FT)-IR and FT-Raman spectra of solid sample of 4-chloro-2-fluorotoluene have been studied[1].

Application

4-Chloro-2-fluorotoluene has been used in the preparation of 4-chloro-2-fluoro-benzylbromide[2].

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

123.8 °F - closed cup

Flash Point(C)

51 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Ramalingam et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 78(1), 429-436 (2010-12-15)
FT-IR (4000-100 cm(-1)) and FT-Raman (4000-100 cm(-1)) spectra of solid sample of 4-chloro-2-fluoro toluene (4Cl2FT) have been recorded using Bruker IFS 66 V spectrometer. Ab initio-HF (HF/6-311++G (d, p)) and DFT (B3LYP/6-311++G and B3PW91/6-311++G (d, p)) calculations have been performed
Synthesis and antitumor activity of platinum (II) complexes containing substituted ethylenediamine ligands.
Brunner H, et al.
European Journal of Medicinal Chemistry, 25(1), 35-44 (1990)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service