Skip to Content
Merck
All Photos(1)

Key Documents

245674

Sigma-Aldrich

(E)-Benzaldehyde oxime

97% (mixture of cis and trans), (Z)-Benzaldehyde oxime ≤6%

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$71.00
50 G
$232.00

$71.00


Available to ship on22 April 2025Details


Request a Bulk Order

Select a Size

Change View
10 G
$71.00
50 G
$232.00

About This Item

Linear Formula:
C6H5CH=NOH
CAS Number:
Molecular Weight:
121.14
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$71.00


Available to ship on22 April 2025Details


Request a Bulk Order

Quality Level

Assay

97% (mixture of cis and trans)

form

solid

refractive index

n20/D 1.591 (lit.)

bp

104 °C/6 mmHg (lit.)

mp

34-36 °C (lit.)

solubility

methanol: 1 g/10 mL

functional group

amine
oxime
phenyl

SMILES string

O\N=C\c1ccccc1

InChI

1S/C7H7NO/c9-8-6-7-4-2-1-3-5-7/h1-6,9H/b8-6+

InChI key

VTWKXBJHBHYJBI-SOFGYWHQSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The reduction of (E)-benzaldehyde oxime, at various pH, was studied[1].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

228.2 °F - closed cup

Flash Point(C)

109 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Moa Andresen Bergström et al.
Journal of medicinal chemistry, 51(8), 2541-2550 (2008-03-29)
Metabolic activation of chemicals (prohaptens) in the skin can cause allergic contact dermatitis. We have explored structure-allergenic activity relationships for seven potential oxime prohaptens using the local lymph node assay and a GSH trapping screen with liver microsomes. The general
On the electrolytic reduction of oximes and semicarbazones in alkaline solution.
Lund H.
Tetrahedron Letters, 9(33), 3651-3654 (1968)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service