Skip to Content
Merck
All Photos(2)

Key Documents

245100

Sigma-Aldrich

2,2,6-Trimethyl-4H-1,3-dioxin-4-one

95%

Synonym(s):

Diketene acetone adduct

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$195.00

$195.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 G
$195.00

About This Item

Empirical Formula (Hill Notation):
C7H10O3
CAS Number:
Molecular Weight:
142.15
Beilstein:
2408
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$195.00


Check Cart for Availability

Request a Bulk Order

Assay

95%

form

liquid

refractive index

n20/D 1.460 (lit.)

bp

~275 °C (lit.)
65-67 °C/2 mmHg (lit.)

mp

12-13 °C (lit.)

solubility

water: insoluble

density

1.07 g/mL at 25 °C (lit.)

functional group

ester
ether
ketal

SMILES string

CC1=CC(=O)OC(C)(C)O1

InChI

1S/C7H10O3/c1-5-4-6(8)10-7(2,3)9-5/h4H,1-3H3

InChI key

XFRBXZCBOYNMJP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,2,6-Trimethyl-4H-1,3-dioxin-4-one is used as a building block in organic synthesis and serves as a direct precursor to β-dicarbonyl compounds[1]

Application

2,2,6-Trimethyl-4H-1,3-dioxin-4-one was used in the synthesis of acetylketene by flash pyrolysis[2].

Other Notes

remainder acetone

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

57.2 °F - closed cup

Flash Point(C)

14 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

2, 2, 6-Trimethyl-4H-1, 3-dioxin-4-one
M Seyyedhamzeh, et al.
Synlett, 23, 1556-1557 (2012)
David M. Birney et al.
The Journal of organic chemistry, 62(21), 7114-7120 (2001-10-24)
Acetylketene (1) was generated by flash pyrolysis of 2,2,6-trimethyl-4H-1,3-dioxin-4-one (6). The selectivities of 1 toward a number of representative functional groups were measured for the first time in a series of competitive trapping reactions. The trend in reactivities toward 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service