Skip to Content
Merck
All Photos(1)

Key Documents

244341

Sigma-Aldrich

2-Butyn-1-ol

98%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$105.00
25 G
$278.00

$105.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$105.00
25 G
$278.00

About This Item

Linear Formula:
CH3C≡CCH2OH
CAS Number:
Molecular Weight:
70.09
Beilstein:
1733676
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$105.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.453 (lit.)

bp

142-143 °C (lit.)

mp

−2.2 °C (lit.)

density

0.937 g/mL at 25 °C (lit.)

functional group

hydroxyl

SMILES string

CC#CCO

InChI

1S/C4H6O/c1-2-3-4-5/h5H,4H2,1H3

InChI key

NEEDEQSZOUAJMU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Butyn-1-ol was used in the synthesis of homocoupling products, [{Ru(P(OCH3)3)2(CH3CN)3}2{μ-SCRCHCH(OCH2CCR)S}](CF3SO3)4[1].
Synthesis of macrocyclic ethers via metathesis reactions of cyclohexanes and cyclohexenes with allylic and propargylic side chains.†

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

123.8 °F - closed cup

Flash Point(C)

51 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kazuko Matsumoto et al.
Journal of the American Chemical Society, 124(44), 13106-13113 (2002-10-31)
A variety of inter- and intramolecular dehydration was found in the reactions of [[Ru(P(OCH(3))(3))(2)(CH(3)CN)(3)](2)(mu-S(2))](CF(3)SO(3))(4) (1) with hydroxyl substituted alkenes and alkynes. Treatment of 1 with allyl alcohol gave a C(3)S(2) five-membered ring complex, [[Ru(P(OCH(3))(3))(2)(CH(3)CN)(3)](2)[mu-SCH(2)CH(2)CH(OCH(2)CH=CH(2))S]](CF(3)SO(3))(4) (2), via C-S bond formation after
European Journal of Organic Chemistry, 3727-3727 (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service