242152
Tetrahydropapaveroline hydrobromide
98%
Synonym(s):
Norlaudanosoline hydrobromide
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C16H17NO4 · HBr
CAS Number:
Molecular Weight:
368.22
MDL number:
UNSPSC Code:
12352103
Assay
98%
mp
277 °C (dec.) (lit.)
SMILES string
Br.Oc1ccc(CC2NCCc3cc(O)c(O)cc23)cc1O
Looking for similar products? Visit Product Comparison Guide
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service