240818
Melamine
≥99%
Synonym(s):
2,4,6-Triamino-1,3,5-triazine, sym-Triaminotriazine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C3H6N6
CAS Number:
Molecular Weight:
126.12
Beilstein:
124341
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Recommended Products
Assay
≥99%
mp
>300 °C (lit.)
SMILES string
Nc1nc(N)nc(N)n1
InChI
1S/C3H6N6/c4-1-7-2(5)9-3(6)8-1/h(H6,4,5,6,7,8,9)
InChI key
JDSHMPZPIAZGSV-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
replaced by
Product No.
Description
Pricing
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Carc. 2 - Repr. 2 - STOT RE 2
Target Organs
Urinary tract
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
572.0 °F - closed cup
Flash Point(C)
300 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service