Skip to Content
Merck
All Photos(3)

Key Documents

239380

Sigma-Aldrich

2-Formylbenzenesulfonic acid sodium salt hydrate

technical grade, 75%

Synonym(s):

Benzaldehyde-2-sulfonic acid sodium salt

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$117.00
500 G
$350.00

$117.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 G
$117.00
500 G
$350.00

About This Item

Linear Formula:
HCOC6H4SO3Na · xH2O
CAS Number:
Molecular Weight:
208.17 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$117.00


Check Cart for Availability

Request a Bulk Order

grade

technical grade

Assay

75%

form

solid

functional group

aldehyde
sulfonic acid

SMILES string

O.[Na+].[O-]S(=O)(=O)c1ccccc1C=O

InChI

1S/C7H6O4S.Na.H2O/c8-5-6-3-1-2-4-7(6)12(9,10)11;;/h1-5H,(H,9,10,11);;1H2/q;+1;/p-1

InChI key

MGFBXWRKSPLCAK-UHFFFAOYSA-M

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Multi-Responsive Protein Nanocarriers from Anionic Dynamic Covalent Copolymer.
Polym. Chem. (2014)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service