Skip to Content
Merck
All Photos(1)

Key Documents

239178

Sigma-Aldrich

Tri-Chek buffer capsule set

sufficient powder in each capsule to make 100 mL of buffer solution

Sign Into View Organizational & Contract Pricing

Select a Size

1 SET
$150.00

$150.00


Available to ship on16 April 2025Details



Select a Size

Change View
1 SET
$150.00

About This Item

UNSPSC Code:
41100000
NACRES:
NB.79

$150.00


Available to ship on16 April 2025Details


Related Categories

General description

Powder in capsules for preparing standard buffer solutions at pH 4.00, 7.00 and 10.00. There is sufficient powder in each capsule to make 100 mL of buffer solution.

Legal Information

Tri-Chek is a trademark of Micro Essential Laboratory, Inc.

Kit Components Only

Product No.
Description

  • Hydrion color key buffer 1 vial

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service