Skip to Content
Merck
All Photos(1)

Key Documents

23908

Sigma-Aldrich

Boc-Pro-PAM resin

extent of labeling: ~0.7 mmol/g loading

Synonym(s):

Boc-L-proline bound to PAM resin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.22

extent of labeling

~0.7 mmol/g loading

particle size

100-200 mesh

application(s)

peptide synthesis

storage temp.

2-8°C

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service