Skip to Content
Merck
All Photos(1)

Key Documents

238481

Sigma-Aldrich

Formaldoxime trimer hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$127.00

$127.00


Available to ship on22 April 2025Details


Request a Bulk Order

Select a Size

Change View
10 G
$127.00

About This Item

Empirical Formula (Hill Notation):
C3H9N3O3 · xHCl
CAS Number:
Molecular Weight:
135.12 (free base basis)
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$127.00


Available to ship on22 April 2025Details


Request a Bulk Order

form

solid

Quality Level

SMILES string

Cl[H].ON1CN(O)CN(O)C1

InChI

1S/C3H9N3O3.ClH/c7-4-1-5(8)3-6(9)2-4;/h7-9H,1-3H2;1H

InChI key

VRHJXINSVWQXEB-UHFFFAOYSA-N

Application

Formaldoxime trimer hydrochloride has been used in the synthesis of isoxazolidines[1].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of iminomethano-dibenzo [a,e] cyclooctenes by transannular formation and cleavage of isoxazolidines.
Carling RW and Leeson PD.
Tetrahedron Letters, 29(52), 6985-6988 (1988)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service