237523
Palladium on barium carbonate
reduced, extent of labeling: 5 wt. % loading
About This Item
Recommended Products
form
solid
reaction suitability
reagent type: catalyst
core: palladium
greener alternative product characteristics
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
extent of labeling
5 wt. % loading
greener alternative category
SMILES string
[Pd++].[Ba++].[O-]C([O-])=O.[O-]C([O-])=O
InChI
1S/2CH2O3.Ba.Pd/c2*2-1(3)4;;/h2*(H2,2,3,4);;/q;;2*+2/p-4
InChI key
NFXIBAPFNPIUGD-UHFFFAOYSA-J
Application
Other Notes
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral
Storage Class Code
4.1B - Flammable solid hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service