Skip to Content
Merck
All Photos(1)

Key Documents

237388

Sigma-Aldrich

mono-Methyl 5-nitroisophthalate

99%

Synonym(s):

Monomethyl 5-nitrobenzene-1,3-dicarboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H3-3-(CO2H)CO2CH3
CAS Number:
Molecular Weight:
225.15
Beilstein:
1469253
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

mp

180-182 °C (lit.)

SMILES string

COC(=O)c1cc(cc(c1)[N+]([O-])=O)C(O)=O

InChI

1S/C9H7NO6/c1-16-9(13)6-2-5(8(11)12)3-7(4-6)10(14)15/h2-4H,1H3,(H,11,12)

InChI key

ZCRNIIJXDRYWDU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

mono-Methyl 5-nitroisophthalate has been used as starting reagent in the synthesis of 3-amide-5-[(dipropylamino)carbonyl]benzoic acids.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Steven W Kortum et al.
Bioorganic & medicinal chemistry letters, 17(12), 3378-3383 (2007-04-17)
The design and synthesis of a novel series of potent BACE1 hydroxyethylamine inhibitors. These inhibitors feature hydrogen bonding substituents at the C-5 position of the isophthalamide ring with improved selectivity over cathepsin D.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service