Skip to Content
Merck
All Photos(2)

Key Documents

235741

Sigma-Aldrich

Tetrapropyl orthosilicate

95%

Synonym(s):

Tetrapropoxysilane

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$214.00

$214.00


In StockDetails


Request a Bulk Order

Select a Size

Change View
100 G
$214.00

About This Item

Linear Formula:
(CH3CH2CH2O)4Si
CAS Number:
Molecular Weight:
264.43
Beilstein:
1706115
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

$214.00


In StockDetails


Request a Bulk Order

vapor density

>1 (vs air)

Quality Level

Assay

95%

form

liquid

refractive index

n20/D 1.4 (lit.)

bp

94 °C/5 mmHg (lit.)

density

0.916 g/mL at 25 °C (lit.)

SMILES string

CCCO[Si](OCCC)(OCCC)OCCC

InChI

1S/C12H28O4Si/c1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4/h5-12H2,1-4H3

InChI key

ZQZCOBSUOFHDEE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Silica's versatility spans various industries, including biomedical applications.

Hybrid organic-inorganic sol-gel materials containing silica were first called “ORMOSILs” in 1984.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service