Skip to Content
Merck
All Photos(1)

Key Documents

235571

Sigma-Aldrich

1,1,1-Trichloroethane

ReagentPlus®, ≥99%, contains 0.05% low alkyl epoxide as stabilizer

Synonym(s):

Methylchloroform, ‘Chlorothene’

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl3CCH3
CAS Number:
Molecular Weight:
133.40
Beilstein:
1731614
EC Number:
MDL number:
UNSPSC Code:
12190000
PubChem Substance ID:

vapor density

4.6 (vs air)

vapor pressure

100 mmHg ( 20 °C)

product line

ReagentPlus®

Assay

≥99%

autoignition temp.

998 °F

contains

0.05% low alkyl epoxide as stabilizer

expl. lim.

7.5-15 %, 25 °F

refractive index

n20/D 1.4366 (lit.)

bp

74-76 °C (lit.)

mp

−35 °C (lit.)

density

1.336 g/mL at 20 °C (lit.)

SMILES string

CC(Cl)(Cl)Cl

InChI

1S/C2H3Cl3/c1-2(3,4)5/h1H3

InChI key

UOCLXMDMGBRAIB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Please view www.aldrich.com/epaods regarding the EPA′s request for application information of Ozone Depleting Substances

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Irrit. 2 - Ozone 1 - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mathieu Valcke et al.
Inhalation toxicology, 23(14), 863-877 (2011-11-17)
The objective of this study was to assess the impact of the exposure duration and intensity on the human kinetic adjustment factor (HKAF). A physiologically based pharmacokinetic model was used to compute target dose metrics (i.e. maximum blood concentration (C(max))
Keith L Shelton
The Journal of pharmacology and experimental therapeutics, 333(2), 612-620 (2010-02-04)
The present study examined the involvement of the GABAA, N-methyl-D-aspartate (NMDA), nicotinic acetylcholine, and mu-opioid receptor systems in the transduction of the discriminative stimulus effects of the abused inhalant 1,1,1-trichloroethane (TCE). Sixteen B6SJLF1/J mice were trained to discriminate 10 min
Dylan B Millet et al.
Environmental science & technology, 43(4), 1055-1060 (2009-03-27)
We use recent aircraft measurements of a comprehensive suite of anthropogenic halocarbons, carbon monoxide (CO), and related tracers to place new constraints on North American halocarbon emissions and quantify their global warming potential. Using a chemical transport model (GEOS-Chem) we
Peter J Boogaard et al.
Regulatory toxicology and pharmacology : RTP, 59(1), 125-132 (2010-10-12)
REACH requires health risk management for workers and the general population and introduced the concept of Derived No-Effect Level (DNEL). DNELs must be derived for all substances that are classified as health hazards. As with analogues to other health-risk based
M Bruce MacIver
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology, 34(10), 2296-2304 (2009-06-06)
Abused inhalants are widely used, especially among school-age children and teenagers, and are 'gateway' drugs leading to the abuse of alcohol and other addictive substances. In spite of this widespread use, little is known about the effects produced by inhalants

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service