Skip to Content
Merck
All Photos(1)

Documents

235520

Sigma-Aldrich

3-Methyladipoyl chloride

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCOCH2CH2CH(CH3)CH2COCl
CAS Number:
Molecular Weight:
197.06
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.472 (lit.)

bp

117-119 °C/10 mmHg (lit.)

density

1.217 g/mL at 25 °C (lit.)

SMILES string

CC(CCC(Cl)=O)CC(Cl)=O

InChI

1S/C7H10Cl2O2/c1-5(4-7(9)11)2-3-6(8)10/h5H,2-4H2,1H3

InChI key

YDDUPABDRNKFDU-UHFFFAOYSA-N

Application

3-Methyladipoyl chloride has been used in the preparation of :
  • polyester hydrochlorides
  • polyamides, having potential non-linear optical characteristics

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and characterisation of polyamides and polyimides from amino/methylene bis [benzenamine] styryl pyridines.
Peesapati V, et al.
Polymer International, 43(1), 8-12 (1997)
Modification of montmorillonite by cationic polyesters.
Huskic M, et al.
Applied Clay Science, 43(3), 420-424 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service