Skip to Content
Merck
All Photos(1)

Key Documents

234559

Sigma-Aldrich

Ethyl trimethylacetate

99%

Synonym(s):

Ethyl pivalate, Ethyl 2,2-dimethylpropionate

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$131.00

$131.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 G
$131.00

About This Item

Linear Formula:
(CH3)3CCOOC2H5
CAS Number:
Molecular Weight:
130.18
Beilstein:
1747617
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$131.00


Check Cart for Availability

Request a Bulk Order

Assay

99%

refractive index

n20/D 1.391 (lit.)

bp

118 °C (lit.)

density

0.856 g/mL at 25 °C (lit.)

functional group

ester

SMILES string

CCOC(=O)C(C)(C)C

InChI

1S/C7H14O2/c1-5-9-6(8)7(2,3)4/h5H2,1-4H3

InChI key

HHEIMYAXCOIQCJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethyl trimethylacetate undergoes condensation with acetophenone catalyzed by phosphazene base to yield β-trimethylsilyloxy ester[1]. NCI(F) and NCI(NH2) mass spectra of a series of ethyl trimethylacetates have been studied[2].

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

59.0 °F - closed cup

Flash Point(C)

15 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fragmentation reactions of some aliphatic esters in the NCl (F-) and NCl (NH2-) mass spectra.
Grutzmacher HF and Grotemeyer B.
Org. Mass Spectrom., 19(3), 135-142 (1984)
Koji Kobayashi et al.
Chemical communications (Cambridge, England), (29)(29), 3128-3130 (2006-07-21)
The t-Bu-P4 base was found to be an excellent catalyst for the condensation of trimethylsilylacetate or trimethylacetonitrile with carbonyl compounds to form functionalized alkenes and beta-enaminoesters were also synthesized by the condensation with formanilides.
E Akaho et al.
Journal of pharmaceutical sciences, 70(11), 1225-1228 (1981-11-01)
Interactions of phenolic compounds 4-hexylresorcinol and 3,4-dimethylphenol with esters were studied using hexane-ester cosolvent systems by both phase solubility and partitioning methods. The data obtained by the phase solubility method were variable and could not be analyzed by any mathematical
Yulia Kaluzhny et al.
Alternatives to laboratory animals : ATLA, 43(2), 101-127 (2015-05-23)
The 7th Amendment to the EU Cosmetics Directive and the EU REACH Regulation have reinforced the need for in vitro ocular test methods. Validated in vitro ocular toxicity tests that can predict the human response to chemicals, cosmetics and other

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service