Skip to Content
Merck
All Photos(2)

Key Documents

234222

Sigma-Aldrich

Diacetin

technical grade, 50%

Synonym(s):

Glycerol diacetate

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
$48.80
1 L
$160.00

$48.80


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 ML
$48.80
1 L
$160.00

About This Item

Linear Formula:
(CH3COOCH2)2CHOH
CAS Number:
Molecular Weight:
176.17
Beilstein:
1706903
EC Number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$48.80


Check Cart for Availability

Request a Bulk Order

grade

technical grade

Quality Level

vapor density

6.1 (vs air)

vapor pressure

<1 mmHg ( 20 °C)

form

liquid

concentration

50%

refractive index

n20/D 1.440 (lit.)

solubility

alcohol: soluble(lit.)
benzene: soluble(lit.)
carbon disulfide: insoluble (practically)(lit.)
diethyl ether: soluble(lit.)
water: soluble(lit.)

density

1.17 g/mL at 25 °C (lit.)

SMILES string

CC(=O)OCC(O)COC(C)=O.CC(=O)OCC(CO)OC(C)=O

InChI

1S/2C7H12O5/c1-5(8)11-3-7(10)4-12-6(2)9;1-5(9)11-4-7(3-8)12-6(2)10/h7,10H,3-4H2,1-2H3;7-8H,3-4H2,1-2H3

InChI key

TWSUHSVPDUPKDH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Diacetin is widely used as plasticizer[1].

Application

Diacetin has been used to design and evaluate gliclazide push-pull osmotic pump (PPOP) coated with aqueous colloidal polymer dispersions[1].

Other Notes

Contains 1,2-diacetin, 1,3-diacetin and triacetin

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Xin Tang et al.
Drug development and industrial pharmacy, 39(1), 67-76 (2012-02-22)
The objective of present work was to design and evaluate gliclazide push-pull osmotic pump (PPOP) coated with aqueous colloidal polymer dispersions-Eudragit(®) RL 30D and Eudragit(®) RS 30D. The influence of diacetin, diethyl phthalate (DEP), dibutyl sebacate (DBS) and triethyl citrate
Miguel Castañeda-Zárate et al.
Current biology : CB, 31(1), 238-246 (2020-11-07)
During the evolutionary history of flowering plants, transitions between pollinator groups (pollinator shifts) have been frequent,1 and contributed to the spectacular radiation of angiosperms.2 Although the evolution of floral traits during pollinator shifts has been studied in real time under
Irmgard Schäffler et al.
Scientific reports, 5, 12779-12779 (2015-08-08)
The interaction between floral oil secreting plants and oil-collecting bees is one of the most specialized of all pollination mutualisms. Yet, the specific stimuli used by the bees to locate their host flowers have remained elusive. This study identifies diacetin

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service