Skip to Content
Merck
All Photos(1)

Key Documents

233978

Sigma-Aldrich

Sodium oleate

98%

Synonym(s):

cis-9-Octadecenoic acid sodium salt, Oleic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)7CH=CH(CH2)7COONa
CAS Number:
Molecular Weight:
304.44
Beilstein:
4046357
EC Number:
MDL number:
UNSPSC Code:
12352103

Assay

98%

mp

232-235 °C (lit.)

SMILES string

[Na+].CCCCCCCC\C=C/CCCCCCCC([O-])=O

InChI

1S/C18H34O2.Na/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20;/h9-10H,2-8,11-17H2,1H3,(H,19,20);/q;+1/p-1/b10-9-;

InChI key

BCKXLBQYZLBQEK-KVVVOXFISA-M

Looking for similar products? Visit Product Comparison Guide

Application

Oleic acid is used to investigate effects on apolipoprotein B synthesis, degradation and secretion in cultured cells.

Biochem/physiol Actions

Oleic acid increases hepatic secretion of apolipoprotein B100 in hepatocyte cell lines and in mice . Oleic acid inhibits apolipoprotein B100 secretion at higher physiologic doses .

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gisela Bevilacqua Rolfsen Ferreira da Silva et al.
Pharmaceutical development and technology, 23(2), 151-157 (2017-06-02)
Microemulsions (MEs) loaded with methyl dihydrojasmonate (MJ) were developed to improve the aqueous solubility of this drug. The composition of the formulations ranged according to the oil/surfactant ratio (O/S). The MEs were characterized according to diameter of droplets, X-ray diffraction
Rie Ouchi et al.
Cell metabolism, 30(2), 374-384 (2019-06-04)
Human organoid systems recapitulate in vivo organ architecture yet fail to capture complex pathologies such as inflammation and fibrosis. Here, using 11 different healthy and diseased pluripotent stem cell lines, we developed a reproducible method to derive multi-cellular human liver organoids

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service