Skip to Content
Merck
All Photos(1)

Key Documents

232505

Sigma-Aldrich

1,2-Dihydroxynaphthalene

technical grade

Synonym(s):

1,2-Naphthalenediol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H6(OH)2
CAS Number:
Molecular Weight:
160.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

mp

101-103 °C (lit.)

SMILES string

Oc1ccc2ccccc2c1O

InChI

1S/C10H8O2/c11-9-6-5-7-3-1-2-4-8(7)10(9)12/h1-6,11-12H

InChI key

NXPPAOGUKPJVDI-UHFFFAOYSA-N

General description

1,2-Dihydroxynaphthalene is the most sensitive biomarker for an internal exposure to naphthalene in humans.

Application

1,2-Dihydroxynaphthalene was used as substrate to investigate the rate of oxygen consumption by washed cell suspensions of Corynebacterium sp. strain C125 grown on o-xylene, tetralin or succinate.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Huilan Fu et al.
Molecular plant pathology, 21(10), 1337-1352 (2020-08-11)
The basal transcription factor II H (TFIIH) is a multicomponent complex. In the present study, we characterized a TFIIH subunit Tfb5 by analysing loss- and gain-of-function mutants to gain a better understanding of the molecular mechanisms underlying stress resistance and
Vandana P Swetha et al.
Applied and environmental microbiology, 71(10), 5951-5956 (2005-10-06)
Pseudomonas sp. strains C4, C5, and C6 utilize carbaryl as the sole source of carbon and energy. Identification of 1-naphthol, salicylate, and gentisate in the spent media; whole-cell O2 uptake on 1-naphthol, 1,2-dihydroxynaphthalene, salicylaldehyde, salicylate, and gentisate; and detection of
Angela Patricia B Chua et al.
Veterinary sciences, 7(4) (2020-10-21)
Canine monocytic ehrlichiosis caused by Ehrlichia canis is one of the leading tick-borne diseases of dogs, particularly in tropical countries. A highly sensitive and specific diagnostic method is essential for early detection to facilitate treatment. This study was conducted to
Khalid A Ibrahim et al.
Pakistan journal of pharmaceutical sciences, 30(2(Suppl.)), 579-583 (2017-06-27)
Medicated jelly formulations are patient friendly dosage form for pediatric, geriatric and dysphagic patients. These formulations offer rapid dissolution and absorption of drugs through oral mucosa therefore show the early onset of action. The objective of the study was to
R W Eaton et al.
Journal of bacteriology, 174(23), 7542-7554 (1992-12-01)
The reactions involved in the bacterial metabolism of naphthalene to salicylate have been reinvestigated by using recombinant bacteria carrying genes cloned from plasmid NAH7. When intact cells of Pseudomonas aeruginosa PAO1 carrying DNA fragments encoding the first three enzymes of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service