Skip to Content
Merck
All Photos(1)

Key Documents

23205

Sigma-Aldrich

4-Methoxycarbonylphenyl chloroformate

≥98.5% (AT)

Synonym(s):

Methyl 4-chlorocarbonyloxybenzoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H7ClO4
CAS Number:
Molecular Weight:
214.60
Beilstein:
2111107
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

Assay

≥98.5% (AT)

form

solid

bp

103-106 °C/4 mmHg (lit.)

mp

51-54 °C

application(s)

peptide synthesis

storage temp.

2-8°C

SMILES string

COC(=O)c1ccc(OC(Cl)=O)cc1

InChI

1S/C9H7ClO4/c1-13-8(11)6-2-4-7(5-3-6)14-9(10)12/h2-5H,1H3

InChI key

PPVBQEZMYRAXPV-UHFFFAOYSA-N

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service