Skip to Content
Merck
All Photos(1)

Key Documents

230723

Sigma-Aldrich

Chromium(III) chloride hexahydrate

96%

Synonym(s):

Chromic chloride hexahydrate, Chromium trichloride hexahydrate, Hexaaquachromium(III) chloride

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$51.90
500 G
$150.00

$51.90


In StockDetails


Request a Bulk Order

Select a Size

Change View
100 G
$51.90
500 G
$150.00

About This Item

Linear Formula:
CrCl3 · 6H2O
CAS Number:
Molecular Weight:
266.45
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

$51.90


In StockDetails


Request a Bulk Order

Quality Level

Assay

96%

form

solid

reaction suitability

core: chromium
reagent type: catalyst

density

1.76 g/mL at 25 °C (lit.)

SMILES string

[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].Cl[Cr](Cl)Cl

InChI

1S/3ClH.Cr.6H2O/h3*1H;;6*1H2/q;;;+3;;;;;;/p-3

InChI key

LJAOOBNHPFKCDR-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

General description

Chromium(III) chloride hexahydrate is a hydrated form of chromium(III) chloride commonly used as a precursor for the synthesis of other chromium complexes or as a catalyst in organic synthesis.[1] [2]

Application

Chromium(III) chloride hexahydrate can be used as a:
  • Chromium(III) chloride hexahydrate can be used as a catalyst in the isomerization of glucose into fructose.[1]
  • Precursor in the preparation of chromium(III) oxide nanosheets.[2]

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Met. Corr. 1 - Skin Sens. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Two-Dimensional Amorphous Cr2O3 Modified Metallic Electrodes for Hydrogen Evolution Reaction
Physica Status Solidi RRL: Rapid Research Letters, 13(12), 1900025-1900025 (2019)
Catalytic conversion of glucose in dimethylsulfoxide/water binary mix with chromium trichloride: Role of water on the product distribution
Jia S, et al.
Chemical Engineering Journal, 254, 333-339 (2014)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service