Skip to Content
Merck
All Photos(2)

Key Documents

230006

Sigma-Aldrich

Zinc nitrate hydrate

99.999% trace metals basis

Synonym(s):

Dinitric acid zinc salt hydrate

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$231.00
250 G
$1,480.00

$231.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$231.00
250 G
$1,480.00

About This Item

Linear Formula:
Zn(NO3)2 · xH2O
CAS Number:
Molecular Weight:
189.40 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
form:
powder and chunks

$231.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99.999% trace metals basis

form

powder and chunks

impurities

≤15.0 ppm Trace Metal Analysis

mp

36 °C (lit.)

density

2.065 g/mL at 25 °C (lit.)

SMILES string

O.[Zn++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/2NO3.H2O.Zn/c2*2-1(3)4;;/h;;1H2;/q2*-1;;+2

InChI key

FOSPKRPCLFRZTR-UHFFFAOYSA-N

Application

Starting material for the preparation of a precursor for the photochemical deposition of ZnS.[1] Used to synthesize a new microporous, organically templated zinc phosphate. Such compounds have potential applications as catalysts or in other applications involving microporosity.[2]

Pictograms

Flame over circleExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Ox. Sol. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Harmon, S.B. Sevov, S.C.
Chemistry of Materials, 10, 3020-3020 (1998)
Cheon, J. et al.
Journal of the American Chemical Society, 119, 163-163 (1997)

Articles

Colloidal quantum dots (CQDs) are semiconducting crystals of only a few nanometers (ca. 2–12 nm) coated with ligand/surfactant molecules to help prevent agglomeration.

Questions

1–3 of 3 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  3. Could you kindly confirm, what is the molar ratio of water and anhydrous zinc nitrate in the product no. 230006? Because the value one the webside, decribed by 'x' is not clear enough.

    1 answer
    1. Regrettably, water content is not tested for Product 230006. It is encouraged to dry the material prior to use.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service