Skip to Content
Merck
All Photos(1)

Documents

229768

Sigma-Aldrich

Magnesium acetate tetrahydrate

99.999% trace metals basis

Synonym(s):

Acetic acid magnesium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3COO)2Mg · 4H2O
CAS Number:
Molecular Weight:
214.45
Beilstein:
3730605
EC Number:
MDL number:
UNSPSC Code:
12352300
eCl@ss:
39021909
PubChem Substance ID:

Assay

99.999% trace metals basis

form

solid

reaction suitability

core: magnesium

mp

72-75 °C (lit.)

SMILES string

O.O.O.O.CC(=O)O[Mg]OC(C)=O

InChI

1S/2C2H4O2.Mg.4H2O/c2*1-2(3)4;;;;;/h2*1H3,(H,3,4);;4*1H2/q;;+2;;;;/p-2

InChI key

XKPKPGCRSHFTKM-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yoshihito Kitaoku et al.
Planta, 242(4), 895-907 (2015-05-23)
We first solved the crystal structure of class III catalytic domain of a chitinase from fern (PrChiA-cat), and found a structural difference between PrChiA-cat and hevamine. PrChiA-cat was found to have reduced affinities to chitin oligosaccharides and allosamidin. Plant class
Catarina M S S Neves et al.
Physical chemistry chemical physics : PCCP, 17(47), 32044-32052 (2015-11-18)
Due to scarce available experimental data, as well as due to the absence of predictive models, the influence of salts on the solubility of ionic liquids (ILs) in water is still poorly understood. To this end, this work addresses the
Andrey G Malyutin et al.
ACS applied materials & interfaces, 7(22), 12089-12098 (2015-05-20)
Here we explore the formation of virus-like nanoparticles (VNPs) utilizing 22-24 nm iron oxide nanoparticles (NPs) as cores and proteins derived from viral capsids of brome mosaic virus (BMV) or hepatitis B virus (HBV) as shells. To accomplish that, hydrophobic
Tanja Rozman Peterka et al.
Journal of pharmaceutical and biomedical analysis, 110, 67-75 (2015-03-26)
Tacrolimus is macrolide drug that is widely used as a potent immunosuppressant. In the present work compatibility testing was conducted on physical mixtures of tacrolimus with excipients and on compatibility mixtures prepared by the simulation of manufacturing process used for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service